Catalog Number | Description | Color | Max. RCF | Unit Size |
---|---|---|---|---|
3434PK | 2.0mL Graduated, Nonsterile | Natural | 25,000 x g | Pack of 500 |
3453 | 2.0mL Graduated, Sterile | Natural | 25,000 x g | Case of 5000 |
3453PK | 2.0mL Graduated, Sterile | Natural | 25,000 x g | Pack of 500 |
3434 | 2.0mL Graduated, Nonsterile | Natural | 25,000 x g | Case of 5000 |
3451 | 1.5mL Graduated, Sterile | Natural | 26,000 x g | Case of 5000 |
3451PK | 1.5mL Graduated, Sterile | Natural | 26,000 x g | Pack of 500 |
3446 | 0.6mL Graduated, Nonsterile | Natural | 30,000 x g | Case of 10000 |
3446PK | 0.6mL Graduated, Nonsterile | Natural | 30,000 x g | Pack of 1000 |
3449 | 0.6mL Graduated, Sterile | Natural | 30,000 x g | Case of 10000 |
3449PK also known as 3449 | 0.6mL Graduated, Sterile | Natural | 30,000 x g | Pack of 1000 |
Improve sample recovery by significantly reducing surface binding with Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes. Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. With no additives or coatings, these tubes — available in 0.6mL, 1.5mL and 2.0mL sizes — offer maximum sample recovery by significantly reducing surface binding.
Recommended for:
Daily laboratory usage including centrifugation and short-term sample storage