Nalgene™ Clear PETG Diagnostic Bottles with Closure: Sterile, Bulk Pack
Nalgene™ Clear PETG Diagnostic Bottles with Closure: Sterile, Bulk Pack
Thermo Scientific™

Nalgene™ Clear PETG Diagnostic Bottles with Closure: Sterile, Bulk Pack

Sample, store and ship small-volume, valuable reagents with clear PETG diagnostic bottles that are ideal for use in many sterile, small-volume applications.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
5 mL
10 mL
20 mL
Neck Finish:
20-415 mm
Catalog number 342035-0005
Price (USD)/ Case of 500
668.80
Online exclusive
836.00 
Save 167.20 (20%)
-
Add to cart
Capacity (Metric):
5 mL
Neck Finish:
20-415 mm
Price (USD)/ Case of 500
668.80
Online exclusive
836.00 
Save 167.20 (20%)
Add to cart
Nalgene™ Clear PETG Diagnostic Bottles with Closure: Sterile, Bulk Pack
Catalog number342035-0005
Price (USD)/ Case of 500
668.80
Online exclusive
836.00 
Save 167.20 (20%)
-
Add to cart

Sample, store and ship small-volume, valuable reagents using Thermo Scientific™ Nalgene Clear PETG Diagnostic Bottles. These bottles are ideal for use in test kits and in many sterile, small-volume applications.

Features:
  • Polyethylene Terephthalate (PETG) with excellent clarity for viewing contents
  • Non-pyrogenic and non-cytotoxic
  • Leakproof† ensured when used with the includes lined, white high density polyethylene (HDPE) closure

Ordering Information:

Serum vials are packed in shrink-wrapped trays with closures attached.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity0.16 oz., 5 mL
Capacity (Metric)5 mL
Capacity (English)0.16 oz.
ColorClear
Diameter (Metric) Inner Neck11 mm
MaterialPETG
No. per Case500
No. per Pack100
SterilitySterile
Neck Finish20-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1315 for food and beverage use, USP Class VI.
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)44 mm
Product TypeNarrow Mouth Bottle
Width (English)0.876 in.
Height (English)0.45 in.
Closure LinerWith Liner
Closure MaterialHDPE
Closure Size20-415 mm
Diameter (Metric)22 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeCase of 500