Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays
Thermo Scientific™

Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays

Package biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe in these durable, break-resistant media bottles.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
1 L
60 mL
125 mL
500 mL
500 Expanded
Closure Size:
View all
38-430 mm
Fit for Purpose:
Sterile
Catalog number 342023-1000
Price (USD)/ Case of 24
269.13
Online exclusive
322.00 
Save 52.87 (16%)
-
Add to cart
Capacity (Metric):
1 L
Closure Size:
38-430 mm
Fit for Purpose:
Sterile
Price (USD)/ Case of 24
269.13
Online exclusive
322.00 
Save 52.87 (16%)
Add to cart
Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays
Catalog number342023-1000
Price (USD)/ Case of 24
269.13
Online exclusive
322.00 
Save 52.87 (16%)
-
Add to cart

Thermo Scientific™ Nalgene Square PETG Media Bottles are durable, break-resistant, and feature a septum closure for aseptic syringe dispensing or addition of supplements.

Features:
  • HDPE closure with silicone/PTFE septum provides a PTFE fluid contact surface that is inert and nonstick to ensure fluid integrity.

    • Leakproof†, transparent media bottles
    • Square design saves valuable shelf space
    • Bottles and closures are sterile to 10-6 sterility assurance level

    Ordering Information:

    Packed in shrink-wrapped trays in double-polybagged cartons for cleanroom applications.

    Recommended for:

    Ideal for packaging of biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe.

    Note:

    † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity (Metric)1 L
Capacity (English)33.8 oz.
ColorTransparent
DescriptionWith septum closure
Diameter (English)3.7 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric) Inner Neck28 mm
MaterialPETG
No. per Case24
No. per Pack12
SterilitySterile
Certifications/ComplianceMeet USP Class IV standards. Noncytotoxic.
Closure IncludedYes
GraduatedYes
MouthWide
ShapeSquare
Fit for PurposeSterile
Height (Metric)220 mm
Product TypeSquare Bottle
Height (English)8.66 in.
Closure Size38-430 mm
Diameter (Metric)94 mm
Unit SizeCase of 24