1-Step™ TMB ELISA Substrate Solutions
1-Step™ TMB ELISA Substrate Solutions
Actual product may vary
Thermo Scientific™

1-Step™ TMB ELISA Substrate Solutions

Thermo Scientific™ Pierce TMB ELISA Substrates detect horseradish peroxidase (HRP) activity, yielding a blue color (Amax = 370 nm and 652 nm) that changes to yellow (Amax = 450 nm) upon addition of a sulfuric or phosphoric acid stop solution.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
340241-Step Slow TMB-Elisa Substrate Solution, 250 mL
340221-Step Turbo TMB-ELISA Substrate Solution, 250 mL
340281-Step Ultra TMB-ELISA Substrate Solution, 250 mL
340291-Step Ultra TMB-ELISA Substrate Solution, 1 L
34021TMB Substrate Kit, 400 mL
Catalog number 34024
Price (USD)/ 250 mL
125.65
Online exclusive
131.00 
Save 5.35 (4%)
-
Add to cart
Description:
1-Step Slow TMB-Elisa Substrate Solution, 250 mL
Request bulk or custom format
Price (USD)/ 250 mL
125.65
Online exclusive
131.00 
Save 5.35 (4%)
Add to cart
1-Step™ TMB ELISA Substrate Solutions
Catalog number34024
Price (USD)/ 250 mL
125.65
Online exclusive
131.00 
Save 5.35 (4%)
-
Add to cart

TMB (3,3',5,5' tetramethylbenzidine) is a chromagen that yields a blue color when oxidized with hydrogen peroxide (catalyzed by HRP) with major absorbances at 370 nm and 652 nm. The color then changes to yellow (measured at 450 nm) upon addition of sulfuric or phosphoric acid to stop the reaction. TMB is very sensitive and more quickly oxidized than other HRP substrates, resulting in faster color development. Unlike other commercially available substrates, these products contain no DMF or DMSO.

There are three formulations that differ primarily in their sensitivities. Slow TMB is intermediate in sensitivity, ideal for kinetic readings. The sensitivity of the Turbo TMB compares to that of OPD used at approximately 1 mg/mL. Ultra TMB-ELISA produces the highest signal-to-noise ratio and sensitivity in the picogram range.

Thermo Scientific Pierce 1-Step TMB ELISA Substrates detects horseradish peroxidase (HRP) activity, yielding a blue color (Amax = 370 nm and 652 nm) that changes to yellow (Amax = 450 nm) upon addition of a sulfuric or phosphoric acid stop solution.

Features of 1-Step TMB ELISA Substrates:
• Available in traditional or ready-to-use formulations
• No additional reagents or filtering required
• Non carcinogenic
• Various levels of sensitivity and development speed to suit any assay
• No DMF or DMSO present in the reagents

TMB is the most popular chromogenic substrate for HRP detection in ELISA and is available in several formats. The Pierce 1-Step TMB Substrates are one-component substrates that require no preparation before use. Unlike other commercially available substrates, these products contain no DMF or DMSO.

Thermo Scientific Pierce 1-Step Ultra TMB yields the greatest sensitivity among the TMB substrates, followed by 1-Step Turbo TMB and 1-Step Slow TMB. The sensitivity of the 1-Step Turbo TMB is similar to OPD-based substrates formulated at approximately 1 mg/mL. 1-Step Slow TMB has intermediate sensitivity—more sensitive than ABTS, but less sensitive than o-phenylenediamine dihydrochloride (OPD) or 1-Step Turbo TMB. The 1-Step Slow TMB is an ideal substrate for kinetic studies.

TMB (3,3',5,5'-tetramethylbenzidine) is a chromogen that yields a blue color when oxidized, typically as a result of oxygen radicals produced by the hydrolysis of hydrogen peroxide by HRP. For kinetic or non-stopped ELISA assays, the TMB chromogen has maximal absorbances at 370 nm and 652 nm. The color then changes to yellow with the addition of sulfuric or phosphoric acid with maximum absorbance at 450 nm. A green reaction product may result from partial conversion to the yellow product from the blue intermediate. TMB is very sensitive ELISA substrate and is more quickly oxidized than other HRP substrates, resulting in faster color development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

1-Step Substrates are available in three formulations that vary in sensitivity:

  • 1-Step Slow TMB has intermediate sensitivity – more sensitive than ABTS, but less sensitive than OPD or 1-Step Turbo TMB
  • 1-Step Turbo TMB has sensitivity comparable to OPD used at approximately 1mg/mL
  • 1-Step Ultra TMB-ELISA has the highest signal-to-noise ratio and sensitivity in the picogram range

General References:

  1. Josephy, P.D., et al. (1982). J. Biol. Chem. 257(7), 3669-3675.
  2. Josephy, P.D., et al. (1983). J. Biol. Chem. 258(9), 5561-5569.
  3. Liem, H.H., et al. (1979). Anal. Biochem. 98, 388-393.
Specifications
Description1-Step Slow TMB-Elisa Substrate Solution, 250 mL
Product TypeELISA Substrate
Quantity250 mL
Detection MethodColorimetric
Product Line1-Step™
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase)
Shipping ConditionAmbient
Unit Size250 mL
Contents & Storage
Upon receipt store at 4°C, protected from moisture.