Nalgene™ HDPE Diagnostic Bottles with Closure: Nonsterile, Tray Pack
Nalgene™ HDPE Diagnostic Bottles with Closure: Nonsterile, Tray Pack
Thermo Scientific™

Nalgene™ HDPE Diagnostic Bottles with Closure: Nonsterile, Tray Pack

Perform many small-volume applications with these safe diagnostic bottles that are leakproof when used with Nalgene closures. Ideal for use in test kits, sample storage and reagent shipping.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
View all
13-415 mm
Catalog number 322002-9125
Price (USD)/ Case of 1328
879.75
Online exclusive
1035.00 
Save 155.25 (15%)
-
Add to cart
Capacity (Metric):
4 mL
Neck Finish:
13-415 mm
Price (USD)/ Case of 1328
879.75
Online exclusive
1035.00 
Save 155.25 (15%)
Add to cart
Nalgene™ HDPE Diagnostic Bottles with Closure: Nonsterile, Tray Pack
Catalog number322002-9125
Price (USD)/ Case of 1328
879.75
Online exclusive
1035.00 
Save 155.25 (15%)
-
Add to cart

Perform many small-volume applications with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure. These bottles are ideal for use in test kits, sample storage and shipping of small-volume, valuable reagents. They come packed in rigid trays for easy handling.

Natural High Density Polyethylene Bottles:
  • Nonsterile
  • Translucent
  • Natural polypropylene (PP) closure for excellent chemical resistance
  • Durable, uniform walls that are resistant to splitting or puncturing
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity0.125 oz., 4 mL
Capacity (Metric)4 mL
Capacity (English)0.125 oz.
ColorTranslucent
Diameter (Metric) Inner Neck8 mm
MaterialHDPE
No. per Case1328
No. per Pack332
SterilityNon-sterile
Neck Finish13-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthNarrow
Height (Metric)41 mm
Product TypeSample Container
Width (English)0.62 in.
Height (English)1.61 in.
Diameter (Metric)16 mm
Through the Closure DispensingNo
Unit SizeCase of 1328