Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack

Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals in these light-weight bottles which are leakproof when used with Nalgene closures.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
1000 mL
30 mL
60 mL
125 mL
250 mL
500 mL
Closure Size:
View all
63-415 mm
Catalog number 312187-0032
Price (USD)/ Case of 50
183.60
Online exclusive
204.00 
Save 20.40 (10%)
-
Add to cart
Capacity (Metric):
1000 mL
Closure Size:
63-415 mm
Price (USD)/ Case of 50
183.60
Online exclusive
204.00 
Save 20.40 (10%)
Add to cart
Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Catalog number312187-0032
Price (USD)/ Case of 50
183.60
Online exclusive
204.00 
Save 20.40 (10%)
-
Add to cart

Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals with Thermo Scientific™ Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure. These durable, autoclavable bottles are designed for shipping and storage.

Features:
  • Wide mouth for easy filling and dispensing of powders
  • Injection blow molded for uniform wall thickness and strength
  • Made from high-purity, food-grade resins; no plasticizers, fillers or extenders
  • Polypropylene closures included
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorTranslucent
DescriptionWide-Mouth Packaging Bottles
Diameter (Metric) Inner Neck53 mm
MaterialPPCO
No. per Case50
No. per Pack50
Closure ColorNatural, translucent
Neck Finish63 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
AutoclavableAutoclavable
MouthWide
ShapeRound
Height (Metric)199 mm
Product TypeWide Mouth Packer Bottle
Width (English)3.6 in.
Height (English)7.83 in.
Closure MaterialPolypropylene
Closure Size63-415 mm
Diameter (Metric)91 mm
Closure TypeScrew (Bulk Packed, Unassembled)
Through the Closure DispensingNo
Unit SizeCase of 50