Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure: Bulk Pack
Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure: Bulk Pack

Store and ship light-sensitive reagents with these opaque, amber, HDPE bottles, ideal for diagnostics, biologicals, specialty chemicals, adhesives and veterinary pharmaceuticals.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
125 mL
30 mL
60 mL
250 mL
500 mL
1000 mL
Closure Size:
View all
24-415 mm
Catalog number 312085-0004
Price (USD)/ Case of 500
387.24
Online exclusive
461.00 
Save 73.76 (16%)
-
Add to cart
Capacity (Metric):
125 mL
Closure Size:
24-415 mm
Price (USD)/ Case of 500
387.24
Online exclusive
461.00 
Save 73.76 (16%)
Add to cart
Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure: Bulk Pack
Catalog number312085-0004
Price (USD)/ Case of 500
387.24
Online exclusive
461.00 
Save 73.76 (16%)
-
Add to cart

Store and ship light-sensitive reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high-density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.

Features:
  • Opaque amber HDPE Bottle with opaque amber PP Closure
  • Tough plastic construction
  • Leakproof† when used with Nalgene closures

Recommended for:

Storage and shipping of light-sensitive reagents.

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
ColorOpaque Amber
DescriptionNarrow-Mouth Packaging Bottles
Diameter (Metric) Inner Neck18 mm
MaterialHDPE
No. per Case500
No. per Pack500
Closure ColorOpaque Amber
Neck Finish24 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)101 mm
Product TypePacker Bottle
Width (English)1.99 in.
Height (English)3.97 in.
Closure MaterialPolypropylene
Closure Size24-415 mm
Diameter (Metric)50 mm
Closure TypeScrew (Bulk Packed, Unassembled)
Through the Closure DispensingNo
Unit SizeCase of 500