Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack

Package, ship and store a variety of reagents using these round, narrow-mouth LDPE bottles with closure.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
1 L
30 mL
60 mL
125 mL
250 mL
500 mL
Closure Size:
View all
38-430 mm
Catalog number 312003-0032
Price (USD)/ Case of 50
268.60
Online exclusive
316.00 
Save 47.40 (15%)
-
Add to cart
Capacity (Metric):
1 L
Closure Size:
38-430 mm
Price (USD)/ Case of 50
268.60
Online exclusive
316.00 
Save 47.40 (15%)
Add to cart
Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Catalog number312003-0032
Price (USD)/ Case of 50
268.60
Online exclusive
316.00 
Save 47.40 (15%)
-
Add to cart

Package, ship and store a variety of reagents using Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure, constructed from natural low-density polyethylene.

Key Features
  • Durable and clear
  • Uniform walls that resist splitting and puncturing
  • Leakproof† when used with Nalgene closures
  • Ideal for trace metal analysis — LDPE is low in trace metal content

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1 L
Capacity (English)32 oz.
ColorNatural
DescriptionNarrow-Mouth Boston Round Bottle
Diameter (Metric) Inner Neck27 mm
MaterialLDPE
No. per Case50
No. per Pack50
SterilityNon-sterile
Closure ColorNatural
Neck Finish38 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthNarrow
ShapeRound
Height (Metric)216 mm
Product TypeBoston Round Bottle
Height (English)8.5 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38-430 mm
Diameter (Metric)91 mm
Closure TypeYes (Bulk Packed, Unassembled)
Through the Closure DispensingNo
Unit SizeCase of 50