Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Thermo Scientific™

Nalgene™ PPCO Conical-Bottom Centrifuge Tubes

Simplify measurements using these PPCO conical-bottom centrifuge tubes, which feature molded-in graduations. Translucent PPCO has excellent chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Capacity (Metric)Length (Metric)Unit Size
3103-001516.8 mm15 mL118.6 mmCase of 100
3103-0015PK
also known as 3103-0015
16.8 mm15 mL118.6 mmPack of 10
3103-005028.7 mm50 mL133.1 mmCase of 100
3103-0050PK
also known as 3103-0050
28.7 mm50 mL133.1 mmPack of 10
Catalog number 3103-0015
Price (USD)/ Case of 100
385.00
Online exclusive
731.00 
Save 346.00 (47%)
-
Add to cart
Diameter (Metric):
16.8 mm
Capacity (Metric):
15 mL
Length (Metric):
118.6 mm
Unit Size:
Case of 100
Price (USD)/ Case of 100
385.00
Online exclusive
731.00 
Save 346.00 (47%)
Add to cart
Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Catalog number3103-0015
Price (USD)/ Case of 100
385.00
Online exclusive
731.00 
Save 346.00 (47%)
-
Add to cart

Simplify measurements using Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.

Features:
  • Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents
  • Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (Tubes must be filled at least 80% for proper performance)
  • Conical bottoms concentrate pellet in a small area for easy isolation and retrieval
  • Molded-in graduations last the life of the tube
  • Last longer than polycarbonate tubes under conditions of repeated autoclaving
  • Tube are both autoclavable and graduated
  • Autoclavable

Ordering Information:

Supplied without closure; friction fit closures available. For 15mL use DS3111-00017; for 50mL use DS3111-0029.

Recycling Information: 
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.

Specifications
Capacity (Metric)15 mL
MaterialPolypropylene Copolymer
No. per Case100
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeConical
Length (Metric)118.6 mm
Bottom ShapeConical
Writing AreaNo
Diameter (Metric)16.8 mm
GraduationsMolded-in
Product TypeCentrifuge Tube
Max. RCF6000 x g
Unit SizeCase of 100