Thermo Scientific™ NeutrAvidin Protein-HRP is a specially-prepared form of avidin that decreases background in biotin-binding conjugated to peroxidase for substrateRead more
Have Questions?
Catalog number 31001
Price (USD)/ 2 mg
346.65
Online exclusive
353.00
Save 6.35 (2%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 2 mg
346.65
Online exclusive
353.00
Save 6.35 (2%)
Add to cart
NeutrAvidin Protein, HRP
Catalog number31001
Price (USD)/ 2 mg
346.65
Online exclusive
353.00
Save 6.35 (2%)
-
Add to cart
Thermo Scientific™ NeutrAvidin Protein-HRP is a specially-prepared form of avidin that decreases background in biotin-binding conjugated to peroxidase for substrate based detection.
Features include: • Near-neutral isoelectric point—pI = 6.3, more neutral than native avidin • Nearly devoid of glycosylation—decreased possibility of lectin binding compared to native avidin • No RYD recognition sequence—no known off-target binding domains like streptavidin • Affordable—significantly less expensive than streptavidin
NeutrAvidin Protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. Purified and conjugated forms of NeutrAvidin Protein provide exceptional performance in western blot, ELISA, and IHC applications that require biotin-binding probes. Assay specificity, sensitivity, and signal-to-noise ratios with NeutrAvidin Protein are generally equivalent or better than with the significantly more expensive streptavidin.
Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles, and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin-binding proteins. The specific activity for biotin binding is approximately 14μg/mg of protein, which is near the theoretical maximum activity.
Recommended applications: ELISA (assay dependent), immunohistochemistry (assay dependent), in situ hybridization (ISH) (assay dependent), western blot (assay dependent)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateHRP
Quantity2 mg
FormSolid
Protein FamilyAvidin
Target MoleculeBiotin
Label or DyeHRP (Horseradish Peroxidase)
Unit Size2 mg
Contents & Storage
Upon receipt store product 4°C. Product is shipped at ambient temperature.