Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Thermo Scientific™

Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes

Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Membrane)Volume (Metric)Pore SizeFits Bottle Neck Size (Metric)
290-4520SFCA150 mL0.2 μm45mm
596-3320PES150 mL0.2 μm33mm
596-4520PES150 mL0.2 μm45mm
296-3345PES150 mL0.45 μm33mm
296-4545PES150 mL0.45 μm45mm
595-3320PES500 mL0.2 μm33mm
595-4520PES500 mL0.2 μm45mm
295-3345PES500 mL0.45 μm33mm
295-4545PES500 mL0.45 μm45mm
597-3320PES1000 mL0.2 μm33mm
597-4520PES1000 mL0.2 μm45mm
290-3320SFCA150 mL0.2 μm33mm
290-3345SFCA150 mL0.45 μm33mm
290-4545SFCA150 mL0.45 μm45mm
291-3320SFCA500 mL0.2 μm33mm
291-4520SFCA500 mL0.2 μm45mm
291-3345SFCA500 mL0.45 μm33mm
291-4545SFCA500 mL0.45 μm45mm
292-3320SFCA1000 mL0.2 μm33mm
292-4520SFCA1000 mL0.2 μm45mm
Catalog number 290-4520
Price (USD)/ Case of 12
83.30
Online exclusive
122.00 
Save 38.70 (32%)
-
Add to cart
Material (Membrane):
SFCA
Volume (Metric):
150 mL
Pore Size:
0.2 μm
Fits Bottle Neck Size (Metric):
45mm
Price (USD)/ Case of 12
83.30
Online exclusive
122.00 
Save 38.70 (32%)
Add to cart
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Catalog number290-4520
Price (USD)/ Case of 12
83.30
Online exclusive
122.00 
Save 38.70 (32%)
-
Add to cart

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.

  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
DisposableSingle-use
Filter TypeMembrane Filter
MaterialPolystyrene
Material (Membrane)SFCA
Pore Size0.2 μm
SterilitySterile
Fits Bottle Neck Size (Metric)45mm
No. per Case12
No. per Pack12
TypeBottle Top Filter
Volume (Metric)150 mL
Diameter (Metric)50 mm
Diameter (Metric) Membrane50 mm
Fit for PurposeSterile
Unit SizeCase of 12