UHPLC Filters
UHPLC Filters
Thermo Scientific™
UHPLC Filters
Protects columns without compromising performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
27006UHPLC Direct Connect Filter Holder
22180TS
also known as 22180
2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm
Catalog number 27006
Price (USD)/ Each
333.83
Add to cart
Type:
UHPLC Direct Connect Filter Holder
Price (USD)/ Each
333.83
Add to cart
UHPLC Filters
Catalog number27006
Price (USD)/ Each
333.83
-
Add to cart
Sacrificing performance is not an option with Thermo Scientific™ UHPLC filters that protect columns for Hypersil GOLD™ 1.9μm and Syncronis 1.7μm columns. These UHPLC filters provide a low-volume filter cartridge design, maintain peak shape, and ensure minimal efficiency loss through dispersion.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeUHPLC Direct Connect Filter Holder
Product LineThermo Scientific
Unit SizeEach