96 Well Plate, Non-Treated Surface, No lid, Non-sterile, Pack of 5
96 Well Plate, Non-Treated Surface, No lid, Non-sterile, Pack of 5
Thermo Scientific™
96 Well Plate, Non-Treated Surface, No lid, Non-sterile, Pack of 5
These microplates are optimized for robotics, automated readers and liquid handling systems. Ideal for antibiotic screens, cell-based assays and screening compounds.
Have Questions?
Catalog number 269620
Price (USD)/ Case of 60
186.65
Online exclusive
198.00 
Save 11.35 (6%)
-
Add to cart
Price (USD)/ Case of 60
186.65
Online exclusive
198.00 
Save 11.35 (6%)
Add to cart
96 Well Plate, Non-Treated Surface, No lid, Non-sterile, Pack of 5
Catalog number269620
Price (USD)/ Case of 60
186.65
Online exclusive
198.00 
Save 11.35 (6%)
-
Add to cart
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWithout Lid
Description96F well plate, Non-Treated, clear, without lid, Non-sterile (Pack of 5, Case of 60)
Array8 x 12
Packaging5/Pk., 60/Cs.
Product LineNunc
ColorClear
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
No. per Case60
No. per Pack5
Unit SizeCase of 60