Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWith Lid
Culture Area0.35 cm2/well
Array8 x 12
Packaging10 Pk., 160 Cases
Product LineNunc
ColorClear
Product TypeMicroplate
Skirt StyleLow Flange
MaterialPolystyrene
Volume (Metric) Well400 μL
No. of Wells96
SterilitySterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Green FeaturesLess waste, fewer resources used
No. per Case160 Microplates
No. per Pack10
Unit SizeCase of 160