Catalog Number | Color | Sterility | Volume (Metric) Well | Fit for Purpose |
---|---|---|---|---|
267462 | White | Non-sterile | 55 μL | Certified Quality |
267460 | Natural | Sterile | 55 μL | Certified Quality |
267461 | Black | Non-sterile | 55 μL | Certified Quality |
Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.