Hypersil GOLD™ PEI HILIC HPLC Columns
Hypersil GOLD™ PEI HILIC HPLC Columns
Thermo Scientific™

Hypersil GOLD™ PEI HILIC HPLC Columns

Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Hypersil GOLD PEI HILIC LC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
26503-01210110 mm2.1 mm3 μm
26505-05213050 mm2.1 mm5 μm
26505-102130100 mm2.1 mm5 μm
26505-152130150 mm2.1 mm5 μm
26505-154630150 mm4.6 mm5 μm
26505-254630250 mm4.6 mm5 μm
26502-152130150 mm2.1 mm1.9 μm
26502-102130100 mm2.1 mm1.9 μm
26502-05213050 mm2.1 mm1.9 μm
26503-154630150 mm4.6 mm3 μm
26503-153030150 mm3 mm3 μm
26503-152130150 mm2.1 mm3 μm
26503-151030150 mm1 mm3 μm
26503-103030100 mm3 mm3 μm
26503-102130100 mm2.1 mm3 μm
26503-05213050 mm2.1 mm3 μm
26505-103030100 mm3 mm5 μm
26503-01100110 mm1 mm3 μm
26505-01400110 mm4 mm5 μm
Catalog number 26503-012101
Price (USD)/ Each
-
Length (Metric):
10 mm
Diameter (Metric):
2.1 mm
Particle Size:
3 μm
Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Thermo Scientific™ Hypersil GOLD™ PEI HILIC (hydrophilic interaction liquid chromatography) LC Columns. These columns deliver straightforward separation without the need for sample derivatization or ion-pair reagents. Featuring outstanding peak shape and selectivity, the columns provide improved sensitivity for MS detection. With 1.9, 3 and 5μm particle sizes, Hypersil GOLD PEI HILIC Columns are ideal for applications within drug discovery, food testing and environmental analysis.
About the Hypersil GOLD Family of HPLC Columns:

Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5μm particle sizes for all your separation needs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatGuard Cartridge, 4 Pk.
Diameter (Metric)2.1 mm
For Use WithUniguard Direct-Connection and Stand Alone Holder
Length (Metric)10 mm
Max. Pressure5800 psi (400 bar)
Particle Size3 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSilica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeGuard Cartridge
Unit SizeEach