Nunc™ Microplate Lids
Nunc™ Microplate Lids
Thermo Scientific™

Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate CompatibilitySterilityHeight (Metric)
26462396-well PlateSterile7.9 mm
250002All MicroplatesSterile8.34 mm
250003All MicroplatesNon-sterile8.34 mm
250005All MicroplatesNon-sterile8.34 mm
253623All MicroplatesNon-sterile4.6 mm
253624All MicroplatesSterile4.6 mm
26333996-well PlateNon-sterile7.9 mm
26412296-well PlateSterile7.9 mm
264611384-well PlateSterile7.9 mm
264612384-well PlateNon-sterile7.9 mm
264616384-well PlateSterile7.9 mm
Catalog number 264623
Price (USD)/ Case of 60
168.15
Online exclusive
177.00 
Save 8.85 (5%)
-
Add to cart
Plate Compatibility:
96-well Plate
Sterility:
Sterile
Height (Metric):
7.9 mm
Price (USD)/ Case of 60
168.15
Online exclusive
177.00 
Save 8.85 (5%)
Add to cart
Nunc™ Microplate Lids
Catalog number264623
Price (USD)/ Case of 60
168.15
Online exclusive
177.00 
Save 8.85 (5%)
-
Add to cart
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
ColorClear
Dimensions (L x W x H)5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
MaterialPolystyrene
SterilitySterile
No. of Wells96
Height (Metric)7.9 mm
Length (English)5 in.
Length (Metric)127 mm
Condensation RingsYes
Height (English)0.3 in.
No. per Case60
No. per Pack20
Dimensions (L x W)5 x 3.35 in. (127 x 85 mm)
Evaporation BarrierYes
Width (English)3.35 in.
Width (Metric)85 mm
Product TypePlate Lid
Product LineNunc
AdhesiveNone
Temperature Range (Metric)-20°C to +80°C
Well ShapeAll well shapes
Plate Compatibility96-well Plate
Cut-off CornersLong Side
Unit SizeCase of 60