Catalog Number | Well Shape | Sterility | Fit for Purpose |
---|---|---|---|
264573 | Square, Round Bottom (U-well) | Non-sterile | Certified Quality |
264574 | Square, Round Bottom (U-well) | Sterile | Certified Quality |
264575 | Square, Conical Bottom (V-well) | Non-sterile | Certified Quality |
264576 | Square, Round Bottom (U-well) | Non-sterile | Certified Quality |
264579 | Square, Round Bottom (U-well) | Non-sterile | Certified Quality |
4309 | Square, Conical Bottom (V-well) | Sterile | Certified Quality |
4308 | Square, Conical Bottom (V-well) | Non-sterile | Certified Quality |
4312 | Square-Rounded | Non-sterile | Certified Quality |
Minimize costs when storing compounds for small volume applications. Thermo Scientific™ 384-Well Conical Bottom Polypropylene Plates feature a lower binding capacity so proteins or DNA will not bind, allowing for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors for quick identification of your storage plates.