Produce consistent results with this low-bleed, stable column produces.
Have Questions?
Change view
Catalog Number
Particle Size
Diameter (Metric)
Length (Metric)
26088-1421
0.25 μm
0.25 mm
30 m
26088-1435
0.25 μm
0.32 mm
30 m
Catalog number 26088-1421
Price (USD)/ Each
995.29
Add to cart
Particle Size:
0.25 μm
Diameter (Metric):
0.25 mm
Length (Metric):
30 m
Price (USD)/ Each
995.29
Add to cart
TraceGOLD™ TG-WaxMS GC Columns with SafeGuard
Catalog number26088-1421
Price (USD)/ Each
995.29
Add to cart
Produce consistent results with this low-bleed, stable column produces. The Thermo Scientific™ TraceGOLD™ TG-WaxMS GC Column with SafeGuard offers extended analytical column life without a compromise in performance.
Engineered for Superior Protection
Same performance and stability profile as the TraceGOLD TG-WaxMS GC Column
Integrated guard on the GC column for prolonged lifetime of the analytical column
No impact upon performance when cutting column during routine maintenance
Applications:
Fatty acid methyl esters (FAMEs)
Flavor compounds and essential oils
Solvents
Xylene isomers
EPA Method 603 for Acrolein/Acrylonitrile
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
Minimizing column bleed extends column lifetime, leading to higher productivity.
Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Recommended Uses:
Aldehydes
Essential oils
Solvents
Xylenes
Alternative Uses:
Acids
Alcohols
Alcohols in beverages
Alditol acetates (sugars)
Amines - Aliphatic
Amines - Aromatic
Similar To:
DB-WAX
DB-WAXetr
HP-Wax
HP-Innowax
SUPELCOWAX™ 10
CP-Wax 52 CB
Stabilwax™
Rtx™-Wax
BP20
ZB-Wax
Optima™ Wax
AT™-Wax
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.