Nunc™ MicroWell™ 96-Well Microplates
Nunc™ MicroWell™ 96-Well Microplates
Thermo Scientific™
Nunc™ MicroWell™ 96-Well Microplates
Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLidSkirt StyleSterilityBarcode
260844Without LidHigh FlangeNon-sterileIncludes Barcode
260887With LidHigh FlangeSterileIncludes Barcode
269787Without LidLow Flange PinchbarSterileNo Barcode
Catalog number 260844
Price (USD)/ Case of 180
1,038.00
-
Add to cart
Lid:
Without Lid
Skirt Style:
High Flange
Sterility:
Non-sterile
Barcode:
Includes Barcode
Price (USD)/ Case of 180
1,038.00
Add to cart
Nunc™ MicroWell™ 96-Well Microplates
Catalog number260844
Price (USD)/ Case of 180
1,038.00
-
Add to cart
Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
  • Made from clear polystyrene with excellent optical properties, ideal for optical measurements.
  • Non-treated plate with hydrophobic surface is suited for suspension cell cultures and a variety of biochemical assays.
  • Cell culture plate received the Thermo Scientific™ Nunclon™ Delta surface treatment that promotes cell adhesion and growth
  • Collagen I and Poly-D-Lysine coated plates are enhanced for finicky adherent cells and biomolecule attachment
  • Different flange designs are compatible with either manual or robotic operations
  • Customized bar coding is available
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
BarcodeIncludes Barcode
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWithout Lid
Description96F well plate, Non-Treated, clear, without lid, Non-sterile, Barcoded (Pack of 10, Case of 180)
Array8 x 12
Packaging10 Pk.
Product LineNunc
ColorClear
Product TypeMicroplate
Skirt StyleHigh Flange
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
No. per Case180
No. per Pack10
Quantity180/Cs.
Unit SizeCase of 180