96 Well Plate, Non-Treated, Surface, No Lid, Non-sterile, Pack of 10
96 Well Plate, Non-Treated, Surface, No Lid, Non-sterile, Pack of 10
Thermo Scientific™
96 Well Plate, Non-Treated, Surface, No Lid, Non-sterile, Pack of 10
Clear polystyrene plates are ideal for optical measurements. Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment.
Have Questions?
Catalog number 260836
Price (USD)/ Case of 180
594.00
-
Add to cart
Price (USD)/ Case of 180
594.00
Add to cart
96 Well Plate, Non-Treated, Surface, No Lid, Non-sterile, Pack of 10
Catalog number260836
Price (USD)/ Case of 180
594.00
-
Add to cart
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWithout Lid
Description96F well plate, Non-Treated, clear, without lid, Non-sterile (Pack of 10, Case of 180)
Array8 x 12
Packaging10 Pk., 180 Case
Product LineNunc
ColorClear
Product TypeMicroplate
MaterialPolystyrene
Volume (Metric) Well400 μL
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
No. per Case180
No. per Pack10
Unit SizeCase of 180