Specifications
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
Stationary PhaseTracePLOT™ TG-BOND S
Max. TemperatureUp to 200°C
For Use With (Application)GC for Non-Polar and Mid-Polar Compounds
Film Thickness10 μm
Length (Metric)15 m
PolarityMid-Polar
Unit SizeEach