Bacto™ TC Lactalbumin Hydrolysate
Bacto™ TC Lactalbumin Hydrolysate
Actual product may vary
Gibco™

Bacto™ TC Lactalbumin Hydrolysate

Bacto TC Lactalbumin Hydrolysate, an animal origin peptone, is the enzymatically hydrolyzed protein portion of milk whey. This supplement isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
259962500 g
25996110 kg
Catalog number 259962
Price (USD)/ 500 g
115.00
-
Add to cart
Quantity:
500 g
Price (USD)/ 500 g
115.00
Add to cart
Bacto™ TC Lactalbumin Hydrolysate
Catalog number259962
Price (USD)/ 500 g
115.00
-
Add to cart
Bacto TC Lactalbumin Hydrolysate, an animal origin peptone, is the enzymatically hydrolyzed protein portion of milk whey. This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Bacto TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.

Specifications
Culture TypeMammalian, Insect, Microbial
FormPowder
Product TypeDehydrated Culture Media
For Use With (Application)Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines
Product LineBacto™
Quantity500 g
SourceAnimal Origin
Unit Size500 g
Contents & Storage
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.