Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format
Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format
Thermo Scientific™

Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format

Thermo Scientific Pierce Sulfo-NHS, No-Weigh Format is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
24510500 mg
A3926910 x 2 mg
245255 g
Catalog number 24510
Price (USD)/ 500 mg
237.65
Online exclusive
252.00 
Save 14.35 (6%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ 500 mg
237.65
Online exclusive
252.00 
Save 14.35 (6%)
Add to cart
Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format
Catalog number24510
Price (USD)/ 500 mg
237.65
Online exclusive
252.00 
Save 14.35 (6%)
-
Add to cart
Thermo Scientific Pierce Sulfo-NHS, No-Weigh Format is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilization methods.

Features of Sulfo-NHS:

• Efficiency of EDC-mediated coupling is increased in the presence of Sulfo-NHS
• Amine-reactive NHS esters or Sulfo-NHS esters can be made with any carboxyl-containing molecule
• Sulfo-NHS derivatives are usually directly water-soluble (can be added directly to physiologic buffers) and membrane-impermable (can be used for cell surface labeling)
• High-purity, crystalline Sulfo-NHS can be used to create high-quality activated derivatives

Sulfo-NHS (N-hydroxysulfosuccinimide) enables control and modification of carbodiimide crosslinking reactions involving activation of carboxylates (—COOH) for conjugation with primary amines (—NH2). Derivatives are easily synthesized by mixing the Sulfo-NHS with a carboxyl-containing molecule and a dehydrating agent such as the carbodiimide EDC (EDAC). The method is the basis for generating many types of protein labeling reagents, including amine-reactive fluorescent dyes, biotin affinity tags and pegylation compounds.

Applications:
• Improve efficiency of EDC coupling reactions
• Convert carboxyls to amine-reactive Sulfo-NHS esters
• Crosslink proteins to carboxyl-coated beads or surfaces more efficiently
• Activate nanoparticles with amine-reactive Sulfo-NHS esters

Specifications for Sulfo-NHS:
We manufacture N-hydroxysulfosuccinimide to the highest possible specifications to produce the most specific bioconjugates, to ensure the integrity of your data and to provide you with the highest degree of consistency. Each lot of Sulfo-NHS is tested to meet the following minimum specifications:

Purity—Greater than 95% by quantitative NMR (the highest standard for crosslinker purity);
average lot purity is greater than 99%
Solubility—Sample dissolves at 2 mg/mL in deionized water to yield a clear, colorless solution
Identity—IR scan shows only peaks characteristic of N-hydroxysulfosuccinimide

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)
NHS (N-hydroxysuccinimide)
EDC (1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine-Carboxyl
CleavableNo
DescriptionSulfo-NHS
Molecular Weight (g/mol)217.13
PEGylatedNo
Cell PermeabilityNo
Shipping ConditionAmbient
Product LinePierce™
Labeling MethodChemical Labeling
Crosslinker TypeHeterobifunctional
Reactive MoietyCarbodiimide
SpacerShort (<10 Å)
FormPowder
Quantity500 mg
SolubilityWater
FormatStandard, Single-use, Premium-grade
Water SolubleYes
Product TypeCrosslinker
Unit Size500 mg
Contents & Storage
Upon receipt store at 4°C.