Nalgene™ Right-to-Understand FEP Safety Wash Bottles with Globally Harmonized System (GHS) Labeling for Harsh Chemicals
Nalgene™ Right-to-Understand FEP Safety Wash Bottles with Globally Harmonized System (GHS) Labeling for Harsh Chemicals
Thermo Scientific™

Nalgene™ Right-to-Understand FEP Safety Wash Bottles with Globally Harmonized System (GHS) Labeling for Harsh Chemicals

Prevent chemical accidents in the lab with these FEP Wash Bottles. Bottles labeled following the Globally Harmonized System (GHS).
Have Questions?
Change viewbuttonViewtableView
For Use With (Application):
Toluene (Methylbenzene)
Methylene Chloride (Dichloromethane)
Xylene
Acetone
n-Hexane
Methyl Ethyl Ketone (2-butanone)
Capacity (Metric):
500 mL
Unit Size:
Case of 4
Pack of 1
Catalog number 2429-0502
Price (USD)/ Case of 4
831.00
Online exclusive
1579.00 
Save 748.00 (47%)
-
Add to cart
For Use With (Application):
Toluene (Methylbenzene)
Capacity (Metric):
500 mL
Unit Size:
Case of 4
Price (USD)/ Case of 4
831.00
Online exclusive
1579.00 
Save 748.00 (47%)
Add to cart
Nalgene™ Right-to-Understand FEP Safety Wash Bottles with Globally Harmonized System (GHS) Labeling for Harsh Chemicals
Catalog number2429-0502
Price (USD)/ Case of 4
831.00
Online exclusive
1579.00 
Save 748.00 (47%)
-
Add to cart

Prevent chemical accidents in the lab with Nalgene™ Right-to-Understand FEP Wash Bottles. Bottles are permanently etched with chemical identification and safe handling information following the Globally Harmonized System (GHS) specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer harsh laboratory chemicals from manufacturers' containers. Six bottles available. For use with methyl ethyl ketone; n-hexane; toluene; xylene; methylene chloride; acetone.

  • Bottles are permanently etched with 100% Globally Harmonized System (GHS) chemical safety information for easy chemical identification at a glance
  • No other system markings are used on the bottles, which eliminates confusion
  • Bottles are made from high quality Nalgene materials for low extractables and long life in the lab
  • Dispensing nozzles are carefully molded to deliver a straight stream for precision dispensing and safety
  • Bottles are not vented
  • Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof service
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Specifications
Capacity500mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionWith stem and draw tube
For Use With (Application)Toluene (Methylbenzene)
MaterialFEP
No. per Case4
No. per Pack1
Closure ColorNatural
GraduationsNo
Certifications/ComplianceCompliant with GHS Labeling Guidelines
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Product TypeSafety Wash Bottle
Closure MaterialETFE
Closure Size28 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 4