Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Thermo Scientific™

Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles

Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Ethyl Alcohol
Methanol
Sodium Hypochlorite
Isopropanol
Distilled Water
Assorted
Acetone
Unit Size:
View all
Case of 24
Pack of 6
Catalog number 2425-0502
Price (USD)/ Case of 24
259.20
Online exclusive
288.00 
Save 28.80 (10%)
-
Add to cart
Capacity (Metric):
500 mL
Chemical Label:
Ethyl Alcohol
Unit Size:
Case of 24
Price (USD)/ Case of 24
259.20
Online exclusive
288.00 
Save 28.80 (10%)
Add to cart
Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Catalog number2425-0502
Price (USD)/ Case of 24
259.20
Online exclusive
288.00 
Save 28.80 (10%)
-
Add to cart

Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.

Features:

  • Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
  • Self-venting, so contents stay in bottle; prevents self-dispensing
  • Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
  • Color bar that corresponds to colored closure to prevent cross-contamination
  • Designed to dispense through the top with a closure and stem molded in one piece for uniform service
  • Tip that can be cut back to increase flow
  • Tight fit of draw tube and closure that prevents separation when squeezing bottle

Ordering Information:

Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.

.

Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorNatural
DescriptionRTK Wash Bottle
MaterialLDPE
No. per Case24
No. per Pack6
Closure ColorNatural
Certifications/ComplianceMeet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)].
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Height (Metric)210.82 mm
Product TypeSafety Wash Bottle
Height (English)8.3 in.
Chemical LabelEthyl Alcohol
Closure MaterialPolypropylene
Closure Size28 mm
Diameter (Metric)72.64 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24