Nalgene™ Wide-Mouth LDPE Wash Bottles
Nalgene™ Wide-Mouth LDPE Wash Bottles
Thermo Scientific™

Nalgene™ Wide-Mouth LDPE Wash Bottles

Pour without sputter with these wide-mouth LDPE wash bottles which feature through-the-top closure and stem molded in one piece.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
500 mL
1000 mL
Closure Size:
View all
53 mm
Unit Size:
View all
Case of 24
Pack of 4
Catalog number 2407-0500
Price (USD)/ Case of 24
204.30
Online exclusive
227.00 
Save 22.70 (10%)
-
Add to cart
Capacity (Metric):
500 mL
Closure Size:
53 mm
Unit Size:
Case of 24
Price (USD)/ Case of 24
204.30
Online exclusive
227.00 
Save 22.70 (10%)
Add to cart
Nalgene™ Wide-Mouth LDPE Wash Bottles
Catalog number2407-0500
Price (USD)/ Case of 24
204.30
Online exclusive
227.00 
Save 22.70 (10%)
-
Add to cart

High quality squeeze bottles with wide mouth for easy filling. Thermo Scientific™ Nalgene™ Wide Mouth LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

  • Polypropylene screw closure/stem with PPCO draw tube
  • Tip can be cut back for increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
.

Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
DescriptionPolypropylene copolymer draw tube
MaterialLDPE
No. per Case24
No. per Pack4
Closure IncludedYes
GraduatedNo
MouthWide
ShapeCylindrical
Height (Metric)216 mm
Product TypeWash Bottle
Height (English)8.5 in.
Closure MaterialPolypropylene
Closure Size53 mm
Diameter (Metric)73.2 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 24