Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Thermo Scientific™

Nalgene™ LDPE Economy Wash Bottles

Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
250 mL
125 mL
500 mL
1000 mL
Closure Size:
View all
24 mm
Unit Size:
View all
Case of 36
Pack of 6
Catalog number 2401-0250
Price (USD)/ Case of 36
207.25
Online exclusive
223.00 
Save 15.75 (7%)
-
Add to cart
Capacity (Metric):
250 mL
Closure Size:
24 mm
Unit Size:
Case of 36
Price (USD)/ Case of 36
207.25
Online exclusive
223.00 
Save 15.75 (7%)
Add to cart
Nalgene™ LDPE Economy Wash Bottles
Catalog number2401-0250
Price (USD)/ Case of 36
207.25
Online exclusive
223.00 
Save 15.75 (7%)
-
Add to cart

High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

Features:
  • Polypropylene screw closure/stem with PPCO draw tube
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials


Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
ColorTranslucent
IncludesStem and draw tube
MaterialLDPE
No. per Case36
No. per Pack6
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeCylindrical
Height (Metric)175 mm
Product TypeEconomy Wash Bottle
Height (English)6.9 in.
Closure MaterialPolypropylene
Closure Size24 mm
Diameter (Metric)61 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCase of 36