96 Well Black Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10
96 Well Black Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10
Thermo Scientific™
96 Well Black Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10
Ensure consistent and reliable results using black and white plates optimized for robotics, plate readers and liquid handling systems.
Have Questions?
Catalog number 237108
Price (USD)/ Case of 180
1,047.65
Online exclusive
1086.00 
Save 38.35 (4%)
-
Add to cart
Price (USD)/ Case of 180
1,047.65
Online exclusive
1086.00 
Save 38.35 (4%)
Add to cart
96 Well Black Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10
Catalog number237108
Price (USD)/ Case of 180
1,047.65
Online exclusive
1086.00 
Save 38.35 (4%)
-
Add to cart
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. Black and white plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics. Available with or without cell culture treatment.
  • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Working volume range: 50 to 250μL/well
  • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
  • Customized bar coding is available
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWithout Lid
Array8 x 12
Packaging10 Pk.
Product LineNunc
ColorBlack
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working50 to 250 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
No. per Case180
No. per Pack10
QuantityCase of 180
Unit SizeCase of 180