Nunc™ F96 MicroWell™ Black and White Polystyrene Plate
Nunc™ F96 MicroWell™ Black and White Polystyrene Plate
Thermo Scientific™

Nunc™ F96 MicroWell™ Black and White Polystyrene Plate

Ensure consistent and reliable results using black and white plates optimized for robotics, plate readers and liquid handling systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSurface TreatmentNo. per Case
236105WhiteNon-treated50
236107WhiteNon-treated160
237105BlackNon-treated50
237107BlackNon-treated160
Catalog number 236105
Price (USD)/ Case of 50
326.65
Online exclusive
343.00 
Save 16.35 (5%)
-
Add to cart
Color:
White
Surface Treatment:
Non-treated
No. per Case:
50
Price (USD)/ Case of 50
326.65
Online exclusive
343.00 
Save 16.35 (5%)
Add to cart
Nunc™ F96 MicroWell™ Black and White Polystyrene Plate
Catalog number236105
Price (USD)/ Case of 50
326.65
Online exclusive
343.00 
Save 16.35 (5%)
-
Add to cart

Ensure consistent and reliable results with minimal crosstalk between wells using the Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. With the low flange pinchbar design, these high quality black and white plates are optimized for robotics, plate readers and liquid handling systems. Available with or without cell culture treatment.

  • Black microplates are recommended for fluorescence measurements with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements with maximum reflection and minimal autoluminescence
  • Working volume range: 50 to 250μL/well
  • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
  • Customized bar coding is available
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWith Lid
Array8 x 12
Packaging50 Cases
ColorWhite
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working50 to 250 mL
SterilitySterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
No. per Case50
No. per Pack1
Quantity50/Cs.
Unit SizeCase of 50