Specifications
Recommended StorageStore at room temperature.
Molecular FormulaC20H10N2Na2O4
MDL NumberMFCD00037500
Quantity25 g
InChI KeyAUPXFICLXPLHBB-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1
IUPAC Namedisodium [2,2'-biquinoline]-4,4'-dicarboxylate
Molecular Weight (g/mol)388.29
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LinePierce
Unit Size25 g