Orion™ 2295 Phosphate Analyzer
Orion™ 2295 Phosphate Analyzer
Thermo Scientific™

Orion™ 2295 Phosphate Analyzer

Our Thermo Scientific™ Orion™ 2295 Phosphate Analyzer is designed to provide continuous and online measurement of phosphates.
Have Questions?
Catalog number 229500
Price (USD)/ Each
-
Request A Quote
Description:
Orion™ 2295 Phosphate Analyzer
Measurement Ranges:
0.2 to 50ppm auto-ranging or user Programmable.
Current Loops:
Two 0/4-20mA – Direct or Reverse Acting (Isolated). Maximum 900ohm load.
Electrical Requirements:
100 to 240VAC 50/60Hz
Phosphates are widely found in industrial water treatments, fertilizers, biological treatments, heat exchangers and other applications. Our Thermo Scientific™ Orion™ 2295 Phosphate Analyzer is designed to provide continuous and online measurement of phosphates. Monitoring and control of phosphate levels reduces the formation of scaling, microorganisms and other effects that decrease equipment efficiency. The optimized design of the Orion 2295 Phosphate Analyzer leads to reduced reagent consumption and a lower cost of ownership. Thermo Scientific™ products are designed to provide fast, accurate and reliable online phosphate detection – optimized for applications that require early detection of dissolved phosphates.
  • Optimized reagent consumption – designed to allow for 45 days between reagent changes
  • Designed to provide accurate and precise measurements – allowing for fine control of the phosphate dosing process
  • Compact size – small footprint supports panel or wall mounting
  • Simplified operation – intuitive menu simplifies navigation
  • Low operating costs – minimal maintenance with low power consumption
  • On-demand mode for non-continuous operation – allows analyzer to be returned to on-line mode rapidly after a plant is started up
Specifications
AccuracyLess than 5% of reading or ±0.5ppm, whichever is greater, from 0.2 to 50ppm.
Communication DetailsRS232 ASCII protocol for data reporting
Current LoopsTwo 0/4-20mA – Direct or Reverse Acting (Isolated). Maximum 900ohm load.
DescriptionOrion™ 2295 Phosphate Analyzer
Detection Limits0.2ppm
Temperature (Metric) Operating5°C to 45°C
TypePhosphate Analyzer
Enclosure RatingNEMA12 (Electronics cabinet), NEMA4X (Fluidics cabinet)
Flow Rate50 to 1000mL/min
Inlet/Outlet1/4 in. O.D. flexible tubing, polypropylene or similar material
Max. Relative Humidity90% at 40°C
Measurement Ranges0.2 to 50ppm auto-ranging or user Programmable.
MethodsOptical absorption at 470nm
Regulatory DetailsSafety CE: EN/IEC61010-1, cTUVus; EMC CE: EN61326; FCC Part 15 Class A
Relay DetailsFour SPDT, 2A at 240V - Programmable
RepeatabilityLess than ±3% of reading or ±0.1ppm, whichever is greater from 0.2 to 50ppm.
Resolution0.1ppm in all ranges
Response TimeLess than 10 minutes per analysis
Sample Pressure5psi max.
Sample StreamsOne
Sample SupplyContinuous
Suspended SolidsLess than 60μm
Temperature (English) Operating41° to 113°F
Dimensions (L x W x H)27.5 x 16 x 6.4 in. (698 x 406 x 162mm)
Length (English)27.5 in.
Length (Metric)698 mm
Width (English)16 in.
Width (Metric)406 mm
Height (English)6.4 in.
Height (Metric)162 mm
Weight (English)40 lb.
Weight (Metric)18 kg
Electrical Requirements100 to 240VAC 50/60Hz
Voltage100 to 240 VAC
Frequency50/60 Hz
Unit SizeEach