RheoScope Module for the HAAKE™ MARS™ Rheometer
RheoScope Module for the HAAKE™ MARS™ Rheometer
Thermo Scientific™
RheoScope Module for the HAAKE™ MARS™ Rheometer
Speed formulation development, process optimization and product processability through combined rheological measurement and optical analysis
Have Questions?
Catalog number 222-1912
Price (USD)/ Each
-
Request A Quote
Description:
RheoScope Module for HAAKE MARS Rheometer platform
Data Storage:
Up to 30 images per second in HAAKE RheoWin 4 software
Simultaneously measure rheological properties and changes in the microscopic structure of a sample with the Thermo Scientific™ RheoScope Module for the Thermo Scientific™ HAAKE™ MARS™ Rheometer platform. The results allow researchers to speed up formulation development, process optimization and product processability.
The RheoScope Module for the HAAKE Mars Rheometer offers rheological measurements with optical analysis. It simultaneously measures rheological properties and changes in microscopic structure of tested samples. It consists of an optical microscope, digital video camera and temperature control unit used in conjunction with the MARS Rheometer. You can choose from standard and high temperature range versions.
The RheoScope Module for HAAKE MARS Rheometer Features:
  • Compact and full integration with the HAAKE MARS Rheometer platform
  • All measuring modes, including normal force measurement, available with simultaneous recording of microscopic properties
  • Camera and microscope control via supplied software
  • Simultaneous rheological and optical measurements
  • Visualization of data and images in one software package
  • Analysis of structural changes under shear
  • Image analysis software for determining particle sizes, particle size distribution and structure analysis
Specifications
TypeRheoScope Module
CameraBlack-and-white 'progressive' scan CCD camera, with 1024 x 768 pixels, C connector and IEEE 1394 (Firewire) interface
Data StorageUp to 30 images per second in HAAKE RheoWin 4 software
DescriptionRheoScope Module for HAAKE MARS Rheometer platform
Field Depth5 μm (20X lenses)
Lens5X, 10X, 20X and 50X
Light Source150 W, 12 V, Wavelength Range: 380 to 750 nm
Resolution1 μm (20X lenses)
Unit SizeEach