Protects columns without compromising performance.
Catalog Number | Type |
---|---|
22180TS also known as 22180 | 2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm |
27006 | UHPLC Direct Connect Filter Holder |
Sacrificing performance is not an option with Thermo Scientific™ UHPLC filters that protect columns for Hypersil GOLD™ 1.9μm and Syncronis 1.7μm columns. These UHPLC filters provide a low-volume filter cartridge design, maintain peak shape, and ensure minimal efficiency loss through dispersion.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.