Starter Pak No. 3 provides a variety of yeast and soy products, ideal for processes where an animal origin-free mediumRead more
Have Questions?
Catalog number 215368
Price (USD)/ 1 each
319.00
-
Add to cart
Price (USD)/ 1 each
319.00
Add to cart
Starter Pak No. 3
Catalog number215368
Price (USD)/ 1 each
319.00
-
Add to cart
Starter Pak No. 3 provides a variety of yeast and soy products, ideal for processes where an animal origin-free medium is preferred. It provides conveniently packaged, 100-g, non-GMP samples of these peptones that are best suited for human and animal vaccine production and contains: • Bacto Yeast Extract and Gibco Yeast Extract—these peptones contain a mixture of peptides, amino acids, carbohydrates, and vitamins to support optimal growth in microbial species, and are ideal for the production of vaccines • Bacto TC Yeastolate—this peptone is ideal for CHO-based applications of biotherapeutic monoclonal antibodies and recombinant proteins, as well as vaccine applications • Phytone Peptone and Difco Soytone—these soy-based supplements provide a nutritious source of carbohydrates for successful use in microbial fermentation. Blending these soy peptones with yeast peptones has been shown to provide additional benefit to cultures. • Bacto Malt Extract—a water-soluble portion of malted barley, this peptone also provides carbohydrates for a variety of microbial fermentation processes
Specifications
FormPowder
Product TypeDehydrated Culture Media
For Use With (Application)Human and animal vaccine productions
Quantity1 each
SourceAnimal origin-free, yeast and soy
Unit Size1 each
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.