Difco™ Soytone
Difco™ Soytone
Gibco™

Difco™ Soytone

Difco Soytone is an enzymatic digest of soy. With high concentrations of vitamins and carbohydrates uniquely present in soy, thisRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
212488500 g
21248910 kg
Catalog number 212488
Price (USD)/ 500 g
132.00
-
Add to cart
Quantity:
500 g
Price (USD)/ 500 g
132.00
Add to cart
Difco™ Soytone
Catalog number212488
Price (USD)/ 500 g
132.00
-
Add to cart
Difco Soytone is an enzymatic digest of soy. With high concentrations of vitamins and carbohydrates uniquely present in soy, this supplement offers an alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO.

Features:
• An enzymatic digest of soy
• High concentrations of vitamins and carbohydrate

Applications of Difco Soytone include:
• An alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO

Specifications
Culture TypeMammalian, Insect, Microbial
FormPowder
Product TypeYeast Supplement
For Use With (Application)Mammalian Cell Culture
Product LineDifco™
Quantity500 g
SourceAnimal origin-free
Unit Size500 g
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.