Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptidesRead more
Have Questions?
Change view
Catalog Number
Quantity
211713
500 g
211709
10 kg
2 Options
Catalog number 211713
Price (USD)/ 500 g
271.00
-
Add to cart
Quantity:
500 g
Price (USD)/ 500 g
271.00
Add to cart
Bacto™ Tryptose
Catalog number211713
Price (USD)/ 500 g
271.00
-
Add to cart
Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.
Features: • Reported as beneficial for cell culture applications • Animal origin (AO) nutritional supplement
Applications of Bacto Tryptose include: • Tryptose is very useful for cultivation of streptococci, pneumococci, meningococci, and other fastidious organisms. • Tryptose Peptone is often used as a biomass enhancer for recombinant Escherichia coli production
Specifications
Certifications/ComplianceLUO
FormPowder
Product TypeDehydrated Culture Media
Shelf Life1825 Days
FormatBottle
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
Product LineBacto™
Quantity500 g
SourceAnimal Origin
Unit Size500 g
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.