Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein. StudiesRead more
Have Questions?
Change view
Catalog Number
Quantity
211693
500 g
212220
2 kg
212230
10 kg
211692
50 kg
Catalog number 211693
Price (USD)/ 500 g
428.00
-
Add to cart
Quantity:
500 g
Price (USD)/ 500 g
428.00
Add to cart
Bacto™ Proteose Peptone No. 3
Catalog number211693
Price (USD)/ 500 g
428.00
-
Add to cart
Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein. Studies of peptic digests of animal tissue prepared under varying digestion parameters demonstrated that no single peptone is the most suitable nitrogen source for every microbiological application. These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone.
• Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins • Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media • Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone. It offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.
We also offer alternatives to Bacto Proteose Peptones for scale-up to production applications, including BiTek Proteose Peptone.
Specifications
Certifications/ComplianceLUO, MUO
Culture TypeMammalian, Insect, Microbial
FormPowder
Product TypeBacterial Media
Shelf Life1825 Days
FormatBottle
For Use With (Application)Enzymatic digest of Protein
Product LineBacto™
Quantity500 g
SourceAnimal Origin
Unit Size500 g
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.