Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins,Read more
Have Questions?
Change view
Catalog Number
Quantity
211681
500 g
211680
10 kg
2 Options
Catalog number 211681
Price (USD)/ 500 g
398.00
-
Add to cart
Quantity:
500 g
Price (USD)/ 500 g
398.00
Add to cart
Bacto™ Neopeptone
Catalog number211681
Price (USD)/ 500 g
398.00
-
Add to cart
Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation. Customers have used Bacto Neopeptone as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A streptococci, as well as Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health applications.
Features: • Enzymatic digest of bovine (milk) and porcine protein • Animal origin nutritional supplement • Contains a wide variety of peptide sizes in combination with vitamins, nucleotides, and minerals
Applications of Bacto Neopeptone include: • Recommended for use in media for detection of fungi • Reported to provide nutrients for support of spirochetes and protozoa • Microbial fermentation • Production of vaccines for animal health applications
Specifications
Certifications/ComplianceLUO
FormPowder
Product TypeNeopeptone
Shelf Life1825 Days
FormatBottle
For Use With (Application)Production of vaccines for animal health applications
Product LineBacto™
Quantity500 g
SourceAnimal origin, bovine and porcine
Unit Size500 g
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.