Catalog Number | Description | Includes |
---|---|---|
2115201 | Orion Dual Star pH/ISE dual channel benchtop meter ammonia & pH kit with Orion ammonia ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand | Dual Star pH/ISE dual channel meter, 9512BNWP Orion ammonia ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 951207 Orion 100 ppm ammonia standard (475 mL), 951211 Orion ammonia ISA ionic strength adjuster (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate |
2115001 | Orion Dual Star pH/ISE dual channel benchtop meter kit with ROSS Ultra glass-body pH electrode, ATC probe, solutions and stand | Dual Star Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 810001 ROSS Storage Solution; 910104, 910107, 910110 pH 4.01, 7.00, 10.01 pH Buffers |
2115202 | Orion Dual Star pH/ISE dual channel benchtop meter chloride & pH kit with Orion chloride ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand | Dual Star pH/ISE dual channel meter, 9617BNWP Orion chloride ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 941708 Orion 1000 ppm chloride standard (475 mL), 940011 Orion chloride ISA ionic strength adjuster (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate |
2115203 | Orion Dual Star pH/ISE dual channel benchtop meter fluoride & pH kit with Orion fluoride ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand | Dual Star pH/ISE dual channel meter, 9609BNWP Orion fluoride ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 040908 Orion 10 ppm fluoride standard premixed with TISAB II (475 mL), 040906 Orion 1 ppm fluoride standard premixed with TISAB II (475 mL), 940909 Orion TISAB II total ionic strength adjustment buffer (3.8 L), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate |
2115204 | Orion Dual Star pH/ISE dual channel benchtop meter nitrate & pH kit with Orion nitrate ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand | Dual Star pH/ISE dual channel meter, 9707BNWP Orion nitrate ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 930707 Orion 100 ppm nitrate standard (475 mL), 930711 Orion nitrate ISA ionic strength adjuster (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate |
2115205 | Orion Dual Star pH/ISE dual channel benchtop meter sodium & pH kit with ROSS sodium ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and stand | Dual Star pH/ISE dual channel meter, 8611BNWP ROSS sodium ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 841108 Orion 1000 ppm sodium standard (475 mL), 841109 Orion KAP sodium standard & ISA solution (475 mL), 841111 Orion sodium ISA ionic strength adjuster (475 mL), 841113 Orion sodium electrode conditioning solution (475 mL), 841101 Orion sodium electrode storage solution (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate |
2115101 | Orion Dual Star pH/ISE dual channel benchtop meter with ROSS Ultra glass-body pH electrode, ATC probe and stand | Dual Star pH/ISE dual channel meter, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate |
2115102 | Orion Dual Star pH/ISE dual channel benchtop meter with ROSS Ultra Triode epoxy-body pH/ATC electrode | Dual Star pH/ISE dual channel meter, 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, universal power adapter, literature, computer interface cable and meter test certificate |
2115000 | Orion Dual Star pH/ISE dual channel benchtop meter with ATC probe and stand | Dual Star pH/ISE dual channel meter, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate |
Double your measurement capabilities with the Thermo Scientific™ Orion™ Dual Star™ pH and ISE Benchtop Meter. Record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements on two available meter channels, ideal for high throughput pH and ion analysis in the lab. Perform up to 6 point pH and ISE calibrations, log up to 1000 data point sets with time/date stamp and easily transfer to a printer or computer. Use the two channels to measure a pH sample and ISE sample, two pH samples or two ISE samples simultaneously or view each channel separately.
The Orion Dual Star pH/ISE dual channel benchtop meter accelerates sample measurement throughput by showing the results of two channels simultaneously on its large, backlit display. Measure pH, ion concentration (ISE), mV, relative mV or ORP with temperature on two channels using the meter’s two BNC, reference pin-tip and ATC temperature inputs. Collect critical measurements using an assortment of read modes and onscreen ready indicator that alerts when readings are stable. The meter’s Smart Stability and Smart Averaging functions automatically compensate for measurement conditions, so the meter displays more accurate results faster.
Warranty: 3-year meter replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.