Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter
Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter
Thermo Scientific™

Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter

Accurately monitor pH, ion concentration, mV, ORP and temperature on two channels with our pH/ISE benchtop meter for advanced lab analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionIncludes
2115000Orion Dual Star pH/ISE dual channel benchtop meter with ATC probe and standDual Star pH/ISE dual channel meter, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate
2115001Orion Dual Star pH/ISE dual channel benchtop meter kit with ROSS Ultra glass-body pH electrode, ATC probe, solutions and standDual Star Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 810001 ROSS Storage Solution; 910104, 910107, 910110 pH 4.01, 7.00, 10.01 pH Buffers
2115201Orion Dual Star pH/ISE dual channel benchtop meter ammonia & pH kit with Orion ammonia ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and standDual Star pH/ISE dual channel meter, 9512BNWP Orion ammonia ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 951207 Orion 100 ppm ammonia standard (475 mL), 951211 Orion ammonia ISA ionic strength adjuster (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate
2115202Orion Dual Star pH/ISE dual channel benchtop meter chloride & pH kit with Orion chloride ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and standDual Star pH/ISE dual channel meter, 9617BNWP Orion chloride ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 941708 Orion 1000 ppm chloride standard (475 mL), 940011 Orion chloride ISA ionic strength adjuster (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate
2115203Orion Dual Star pH/ISE dual channel benchtop meter fluoride & pH kit with Orion fluoride ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and standDual Star pH/ISE dual channel meter, 9609BNWP Orion fluoride ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 040908 Orion 10 ppm fluoride standard premixed with TISAB II (475 mL), 040906 Orion 1 ppm fluoride standard premixed with TISAB II (475 mL), 940909 Orion TISAB II total ionic strength adjustment buffer (3.8 L), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate
2115204Orion Dual Star pH/ISE dual channel benchtop meter nitrate & pH kit with Orion nitrate ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and standDual Star pH/ISE dual channel meter, 9707BNWP Orion nitrate ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 930707 Orion 100 ppm nitrate standard (475 mL), 930711 Orion nitrate ISA ionic strength adjuster (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate
2115205Orion Dual Star pH/ISE dual channel benchtop meter sodium & pH kit with ROSS sodium ion selective electrode, ROSS Sure-Flow glass-body pH electrode, ATC probe, solutions and standDual Star pH/ISE dual channel meter, 8611BNWP ROSS sodium ion selective electrode, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star series stirrer probe, 841108 Orion 1000 ppm sodium standard (475 mL), 841109 Orion KAP sodium standard & ISA solution (475 mL), 841111 Orion sodium ISA ionic strength adjuster (475 mL), 841113 Orion sodium electrode conditioning solution (475 mL), 841101 Orion sodium electrode storage solution (475 mL), 810199 ROSS pH buffer and solution kit, electrode stand, computer interface cable, universal power supply, literature and meter test certificate
2115101Orion Dual Star pH/ISE dual channel benchtop meter with ROSS Ultra glass-body pH electrode, ATC probe and standDual Star pH/ISE dual channel meter, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate
2115102Orion Dual Star pH/ISE dual channel benchtop meter with ROSS Ultra Triode epoxy-body pH/ATC electrodeDual Star pH/ISE dual channel meter, 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, universal power adapter, literature, computer interface cable and meter test certificate
Catalog number 2115000
Price (USD)/ Each
2,007.00
Online exclusive
2230.00 
Save 223.00 (10%)
-
Add to cart
Description:
Orion Dual Star pH/ISE dual channel benchtop meter with ATC probe and stand
Includes:
Dual Star pH/ISE dual channel meter, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate
Price (USD)/ Each
2,007.00
Online exclusive
2230.00 
Save 223.00 (10%)
Add to cart
Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter
Catalog number2115000
Price (USD)/ Each
2,007.00
Online exclusive
2230.00 
Save 223.00 (10%)
-
Add to cart

Double your measurement capabilities with the Thermo Scientific™ Orion™ Dual Star™ pH and ISE Benchtop Meter. Record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements on two available meter channels, ideal for high throughput pH and ion analysis in the lab. Perform up to 6 point pH and ISE calibrations, log up to 1000 data point sets with time/date stamp and easily transfer to a printer or computer. Use the two channels to measure a pH sample and ISE sample, two pH samples or two ISE samples simultaneously or view each channel separately.

The Orion Dual Star pH/ISE dual channel benchtop meter accelerates sample measurement throughput by showing the results of two channels simultaneously on its large, backlit display. Measure pH, ion concentration (ISE), mV, relative mV or ORP with temperature on two channels using the meter’s two BNC, reference pin-tip and ATC temperature inputs. Collect critical measurements using an assortment of read modes and onscreen ready indicator that alerts when readings are stable. The meter’s Smart Stability and Smart Averaging functions automatically compensate for measurement conditions, so the meter displays more accurate results faster.
 

  • View large, individual measurements on a full screen display or quickly toggle to a split screen display to view two measurements at the same time
  • Easy-to-follow onscreen text prompts for calibration and setup menus available in a multi-language user interface with English, Spanish, French, German or Italian options
  • EZ Startup menu facilitates quick and easy first-time meter setup
  • Numeric keypad and menu-specific function keys allow for quick and efficient meter setup and data entry
  • Selectable read modes include Auto-Read, On-Ready, At Timed Intervals, Continuous, Value Change in Measurement and Timed for a variety of measurement collection methods for different analytical methods
  • Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
  • Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank and low-concentration stability options or use incremental methods for single/double known addition and single/double known subtraction
  • Non-volatile meter memory holds up to 1000 data points with time and date stamp
  • Easily transfer data via USB and RS232 to a printer or computer with free Orion Dual Star Com communication software
  • Stir samples with ease using the meter-controlled Orion Star stirrer probe
  • Meter-attachable electrode stand assists with moving and maintaining electrodes
  • IP54-rated housing handles splashes and is wall-mountable to save space

 

Warranty: 3-year meter replacement warranty

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
AC AdapterUniversal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003)
OutputsUSB, RS232
Accuracy (ISE)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.002
Accuracy (Temperature)±0.1°C
Advanced ISE FunctionsSegmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit
Alarm OutputHigh/low limit alarm
Backlight OptionYes, adjustable
CalibrationpH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
Calibration Points1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (pH)1 to 6
Certifications/ComplianceCE, TUV 3-1, FCC Class A
Data Management DetailsTransfer single, range or all data points to printer or computer
Data Points1000 with date and time stamp
DescriptionOrion Dual Star pH/ISE dual channel benchtop meter with ATC probe and stand
Calibration Points (ISE)2 to 6
Channels2
Measurement ModespH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature
Range (ISE)0.0001 to 19,900 in ppm, M, mg/L, % or unit-less
Range (mV)±1999.9 mV
Range (pH)-2.000 to 19.999
Resolution (ISE)0.0001 minimum, 1 to 3 significant digits (user selectable)
Resolution (pH)0.1, 0.01, 0.001
Temperature (Metric) Range-5.0°C to +105.0°C
Display TypeGraphic LCD with Backlight
IncludesDual Star pH/ISE dual channel meter, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate
InputsTwo BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
Isopotential Point7.000 pH (adjustable), 1.000 ISE (adjustable)
KeypadNumeric with menu-specific function keys and dual purpose scroll/shortcut keys
LanguagesEnglish, Spanish, French, Italian and German
Log Function TypeAutomatic data logging with Auto-Read, On Ready, At Timed Intervals, Value Change In Measurement and
MemoryNon-volatile memory preserves data log, calibration log and meter settings
Methods10 per channel with password protection
ModelOrion Dual Star pH/ISE/mV/RmV/ORP/Temperature Dual Channel Benchtop Meter
Power SupplyUniversal AC adapter (included)
Probe TypepH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (includes 927007MD Orion ATC probe)
Range (ORP)±1999.9 mV
Range (Relative mV)±1999.9 mV
Relative Humidity Range5 to 85 %, non-condensing
Resolution (mV)0.1 mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Dimensions (L x W x H)24 x 17 x 9 cm (9.5 x 6.7 x 3.5 in.)
Weight (English)2 lb.
Weight (Metric)0.9 kg
Electrical Requirements90 to 260 VAC 50/60 Hz
Voltage90 to 260 VAC
Unit SizeEach