Orion™ 2111LL Low Level Sodium Analyzer
Orion™ 2111LL Low Level Sodium Analyzer
Actual product may vary
Thermo Scientific™

Orion™ 2111LL Low Level Sodium Analyzer

Protect your turbine against destructive sodium levels, including trace amounts, with the reliable and easy to use Thermo Scientific™ Orion™ 2111LL Low Level Sodium Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
2111LL2111LL Low Level Sodium Analyzer
2111LLEN2111LL Low Level Sodium Analyzer With Enclosure
2111LLENG2111LL Low Level Sodium Analyzer With Enclosure and Grab Sampler
2111LLG2111LL Low Level Sodium Analyzer and Grab Sampler
Catalog number 2111LL
Price (USD)/ Each
-
Request A Quote
Description:
2111LL Low Level Sodium Analyzer
Protect your turbine against destructive sodium levels, including trace amounts, with the reliable and easy to use Thermo Scientific™ Orion™ 2111LL Low Level Sodium Analyzer. Available in several configurations including the analyzer instrument only, or with a protective enclosure.

Features:

  • Resistant to drift with Orion™ Ross™ UltraTM electrode technology
  • Reduces hazardous waste
  • Easy to read backlit display for measurement visibility from a distance
  • No moving parts with the pum-less passive diffusion reagent system
  • Rapid response to changes and ultra low level detection with proprietary flow cell

Ordering information:

  • All low level sodium monitors come with step-by-step instructions for quick installation and start-up.
  • Available in several configurations including the analyzer instrument only or with a protective enclosure.
Specifications
Accuracy±5% or 0.01 ppb, whichever is greater
Accuracy (mV)± (0.5 mV + 0.1%)
Accuracy (Temperature)±0.5°C
Alarm Output3
Analog Outputs2, Galvanically Isolated
CalibrationDouble Known Addition
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
Channels1
Communication DetailsModbus with Additional Module
Current Loops2
Description2111LL Low Level Sodium Analyzer
Temperature (Metric) Operating5°C to 35°C
TypeSodium Analyzer
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
IncludesInstrument Only Package
Inlet Pressure8 to 100psig
Keypad TypeMembrane
Max. Relative Humidity5 to 95%, non-condensing
Measurement Ranges0 to 10ppm
Mounting OptionsWall mount
Outputs0 to 20 mA; 4 to 20 mA
Range0.001ppb to 10ppm
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Resolution1; 2; 3; or 4 significant digits
Response Time95% within 2 minutes of injecting a standard solution
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Sample Temperature5 to 45°C
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Length (English)11 in.
Length (Metric)27 cm
Width (English)17 in.
Width (Metric)45 cm
Height (English)26 in.
Height (Metric)65 cm
Weight (English)50 lb.
Weight (Metric)22.7 g
Voltage85 to 132 VAC; 170 to 264 VAC
Amperage5A
Frequency0 to 60 Hz
Channel 1Sodium
Channel 2Optional ph/ORP or Conductivity
Unit SizeEach