Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Thermo Scientific™

Nalgene™ Square Wide-Mouth PPCO Bottles with Closure

Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO bottles, which are contact-clear, autoclavable and resistant to most acids, alcohols and bases.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
1 L
60 mL
175 mL
250 mL
500 mL
Closure Size:
View all
63 mm
Unit Size:
View all
Case of 24
Pack of 6
Catalog number 2110-0032
Price (USD)/ Case of 24
232.40
Online exclusive
332.00 
Save 99.60 (30%)
-
Add to cart
Capacity (Metric):
1 L
Closure Size:
63 mm
Unit Size:
Case of 24
Price (USD)/ Case of 24
232.40
Online exclusive
332.00 
Save 99.60 (30%)
Add to cart
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Catalog number2110-0032
Price (USD)/ Case of 24
232.40
Online exclusive
332.00 
Save 99.60 (30%)
-
Add to cart

Save valuable lab space with the square design of Thermo Scientific™ Nalgene™ Wide-Mouth Square Polypropylene Copolymer Bottles. Wide-mouth design allows easy cleaning, sample filling and retrieval. Made from only high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Resistant to acids, alcohols and bases
  • Contact-clear bottle provides visible liquid level
  • Square design provides a large area for labeling
  • Linerless polypropylene closures provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable
  • Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials

Note:

Completely disengage threads or remove cap before autoclaving.

.

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1 L
Capacity (English)32 oz.
ColorTranslucent
DescriptionWide-Mouth Square Bottle
Diameter (English)3.7 in.
Diameter (English) Inner Neck2 in.
Diameter (Metric) Inner Neck53 mm
MaterialPPCO
No. per Case24
No. per Pack6
SterilityNon-sterile
Closure ColorNatural, translucent
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthWide
ShapeSquare
Height (Metric)181 mm
Product TypeSquare Wide Mouth Bottle
Width (English)3.72 in.
Height (English)7.1 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size63 mm
Diameter (Metric)95 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCase of 24