Difco Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. Our yeast extracts are derived fromRead more
Have Questions?
Change view
Catalog Number
Quantity
210933
500 g
210941
10 kg
Catalog number 210933
Price (USD)/ 500 g
183.00
-
Add to cart
Quantity:
500 g
Price (USD)/ 500 g
183.00
Add to cart
Difco™ Yeast Extract, low-dusting (LD)
Catalog number210933
Price (USD)/ 500 g
183.00
-
Add to cart
Difco Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. Our yeast extracts are derived from primary grown baker’s yeast. These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications. Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation.
Difco Yeast Extract Low-Dusting (LD) was designed to limit the amount of dust generated compared to other spray-dried yeast extracts. Difco Yeast Extract UF is ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g.
Related yeast extract products: • Bacto Yeast Extract—one of the most complete and versatile fermentation supplements available. It is an important ingredient for the microbiological assay of vitamins. Yeast extract is also of value in the assay of antibiotics. B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract. • Gibco Yeast Extract—developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications. • Bacto Yeast Extract, Technical—developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
Specifications
Certifications/ComplianceLUO
Cell TypeBacteria
Culture TypeMammalian, Insect, Microbial
FormPowder
Product TypeDehydrated Culture Media
Shelf Life730 Days
FormatBottle
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
Product LineDifco™
Quantity500 g
SourceYeast, animal origin-free
Unit Size500 g
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.