Nalgene™ Square PETG Media Bottles with Closure
Nalgene™ Square PETG Media Bottles with Closure
Thermo Scientific™

Nalgene™ Square PETG Media Bottles with Closure

Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure, ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
125 mL
30 mL
60 mL
250 mL
500 mL
1 L
2000 mL
Catalog number 2019-0125
Price (USD)/ Case of 48
271.00
-
Add to cart
Capacity (Metric):
125 mL
Price (USD)/ Case of 48
271.00
Add to cart
Nalgene™ Square PETG Media Bottles with Closure
Catalog number2019-0125
Price (USD)/ Case of 48
271.00
-
Add to cart

Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure. Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates, and also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.

Features:

  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Suitable for frozen storage down to -40°C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Sterile and non-pyrogenic
  • Available in low particulate format
  • Supported by extensive validation documentation

Ordering Information:

  • Packed in shrink-wrapped trays

Note:

  • 2 L size has molded-in handgrips

Warranty:

  • 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
Capacity (Metric)125 mL
Capacity (English)4 oz.
ColorClear
Diameter (English)2.13 in.
Diameter (English) Inner Neck1.10 in.
Diameter (Metric) Inner Neck28 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage
MaterialPETG
No. per Case48/Cs.
No. per Pack24/Pk.
SterilitySterile
Temperature Range (Metric)-70°C to 70°C
Certifications/ComplianceBottles and closures are certified radiation sterilized (SAL 10-6), non-pyrogenic and non-cytotoxic, and comply with USP Class VI guidelines.
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)110 mm
Product TypeMedia Bottle
Height (English)4.33 in.
Closure MaterialHDPE
Closure Size38-430 mm
Diameter (Metric)54 mm
Unit SizeCase of 48