Nalgene™ Rectangular Amber HDPE Bottles
Nalgene™ Rectangular Amber HDPE Bottles
Thermo Scientific™

Nalgene™ Rectangular Amber HDPE Bottles

Protect light-sensitive liquids or dry materials from exposure with these space-efficient, amber HDPE bottles, featuring sturdy walls and a wide mouth for easy filling.
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2 L
Closure Size:
View all
28 mm
Unit Size:
View all
Case of 72
Pack of 12
Catalog number 2009-0004
Price (USD)/ Case of 72
294.95
Online exclusive
347.00 
Save 52.05 (15%)
-
Add to cart
Capacity (Metric):
125 mL
Closure Size:
28 mm
Unit Size:
Case of 72
Price (USD)/ Case of 72
294.95
Online exclusive
347.00 
Save 52.05 (15%)
Add to cart
Nalgene™ Rectangular Amber HDPE Bottles
Catalog number2009-0004
Price (USD)/ Case of 72
294.95
Online exclusive
347.00 
Save 52.05 (15%)
-
Add to cart

Protect light-sensitive liquids or dry materials from exposure. Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles take up less shelf space than traditional round bottles. These high-density, polyethylene bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Excellent chemical resistance to most acids, bases and alcohols makes this container great for sampling, shipping or storing liquids or dry materials that are sensitive to UV light.

  • Amber polypropylene screw closure
  • Amber HDPE reduces UV light transmissions to protect light-sensitive liquids
  • Meets U.S. Pharmacopoeia 23 light transmission requirements
  • Rectangular shape is great for space-saving use in the freezer to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
.

Recycling Information
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
ColorAmber
DescriptionAmber Wide-Mouth Rectangular Bottle
Diameter (English) Inner Neck0.8 in.
Diameter (Metric) Inner Neck21 mm
MaterialHDPE
Length (Metric)61 mm
No. per Case72
No. per Pack12
SterilityNon-sterile
Width (Metric)38 mm
Closure ColorOpaque Amber
Certifications/ComplianceMeet U.S. Pharmacopeia XXIII requirements for maximum light transmission.
Closure IncludedYes
GraduatedNo
MouthWide
ShapeRectangular
Height (Metric)102 mm
Product TypeWide Mouth Bottle
Height (English)4 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size28 mm
Diameter (Metric)61 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCase of 72