Partisol™ 2000i-D Dichotomous Air Sampler
Partisol™ 2000i-D  Dichotomous Air Sampler
Thermo Scientific™

Partisol™ 2000i-D Dichotomous Air Sampler

Simultaneously collect high quality samples of fine and coarse ambient particulate matter (PM).
Have Questions?
Catalog number 2000ID
Price (USD)/ Each
-
Request A Quote
Description:
Air Sampler

Simultaneously collect high-quality samples of fine and coarse ambient particulate matter (PM) with the Thermo Scientific™ Partisol™ 2000i-D Dichotomous Air Sampler. Developed on the renowned iSeries software platform design, this sampler features USB ports for improved data downloads, and exchanged 47mm diameter sample filters automatically, which can be preset to a user-defined time interval.

Key Features

  • U.S. EPA PM-2.5, PM-10 and PM-Coarse Equivalent Sampler
  • Enhanced user interface and iSeries communication capabilities
  • Improved data downloads
  • Contains an onboard microprocessor operating system and a large internal data storage buffer
  • Low maintence requirements

Additional iSeries features also include:

  • Flash memory for increased data storage
  • Enhanced Ethernet connectivity
  • Remote data access
  • iPort communication software
  • Large interface screen
Specifications
Temperature (English) Operating-40°F to +122°F
Temperature (Metric) Operating-40°C to +50°C
Certifications/ComplianceCE: 89/336/EEC Electromagnetic Compatibility (EMC
Data Points86 days of Interval data (stored every 5 minutes); 32 filter data records, and input data (30 minute default)
DescriptionAir Sampler
Electrical Requirements2.2 A (at 120 VAC); 1.1 A (at 240 VAC)
Filter Holders and MediaAccommodates 47 mm filter in two sample positions (for USEPA PM-10 and PM-2.5 sampling); PTFE 2 μm pore size membrane filter (regulation required) (for USEPA PM-10 reference sampling: Pallflex TX-40, quartz filter and PTFE materials)
Height (English)24 in. (Overall)/32 in. (Stand)
Height (Metric)61 cm (Overall)/82 cm (Stand)
Sample Flow Control and ReportingActivol flow control system using mass flow controller. Controls at rates ranging at 5 volumetric L/min for fine sample and 1.67lpm for coarse sample. The sampler displays the current volumetric flow rate (l/min.).
Weight (English)75 lb.
Weight (Metric)32 kg
Width (English)16 in. (Overall)/42 in. (Stand)
Width (Metric)41 cm (Overall)/ 108 cm (Stand)
Unit SizeEach