Samco™ Exact Volume Transfer Pipettes
Samco™ Exact Volume Transfer Pipettes
Thermo Scientific™
Samco™ Exact Volume Transfer Pipettes
Perform precise dispensing of volumes from 20 to 400 μL by using these exact volume transfer pipettes. Made of shatterproof, nontoxic polyethylene.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)SterilityDescription
191250 μL10.19 cmNon-sterileSamco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case)
784TS
also known as 784
40 μL5.9 cmNon-sterile40μL Exact Volume Transfer Pipette
785TS
also known as 785
60 μL6.5 cmNon-sterile60μL Exact Volume Transfer Pipette
786TS
also known as 786
80 μL7.5 cmNon-sterile80μL Exact Volume Transfer Pipette
787TS
also known as 787
100 μL8.3 cmNon-sterile100μL Exact Volume Transfer Pipette
941200 μL8.89 cmNon-sterile200μL Exact Volume Double Bulb Transfer Pipettes, Non-sterile; 200μL Exact Volume Double Bulb Transfer Pipettes, Non-sterile (500/Pack, 5000/Case)
967TS
also known as 967
400 μL10.59 cmNon-sterile400μL
788150 μL8.0 cmNon-sterile150μL Exact Volume Transfer Pipette
966NL300 μL9.5 cmNon-sterile300μL Exact Volume Transfer Pipette
Catalog number 191
Price (USD)/ Each
-
Request A Quote
Volume (Metric):
250 μL
Length (Metric):
10.19 cm
Sterility:
Non-sterile
Description:
Samco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case)
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
  • Shatterproof, nontoxic polyethylene
  • Nonsterile
  • Low affinity surface
  • Freezer ready
Specifications
DescriptionSamco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case)
Diameter (Metric) Outer3 mm
MaterialPolyethylene
SterilityNon-sterile
Volume (Metric)250 μL
Length (Metric)10.19 cm
Unit SizeEach