CTP
GMP-quality NTPs are available for RNA vaccine and therapeutic development. See details
Inquire about OEM or Commercial Supply version of this product here.
CTP
Invitrogen™

CTP

ATP, CTP, GTP, UTP (nucleoside 5´-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactionsRead more
Have Questions?
Catalog number 18331017
Price (USD)/ 5 μmol
74.00
-
Add to cart
Request bulk or custom format
Price (USD)/ 5 μmol
74.00
Add to cart
CTP
Catalog number18331017
Price (USD)/ 5 μmol
74.00
-
Add to cart

ATP, CTP, GTP, UTP (nucleoside 5´-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases for production of RNA probes, or RNA transcripts for translation in in vitro translation extracts. These nucleotides are supplied as convenient, ready-to-use solutions at a concentration of 10 mM in 1 mM Tris-HCl (pH 7.5).

Related products
10 mM ATP
10 mM GTP
10 mM UTP
10 mM NTP Mix

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling MethodRNA Labeling
Shipping ConditionApproved for shipment on Wet or Dry Ice
Concentration10 mM
Label or DyeUnlabeled
Quantity500 μL
FormLiquid
Unit Size5 μmol
Contents & Storage
10 mM CTP in Tris-HCl (250 μL)

Store at –5 to –30°C.