PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Thermo Scientific™

PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings

Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap™ Neo Trap Cartridge to your HPLC system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionLength (Metric)For Use With
174501PEEK Tubing with nanoViper™ Fittings100 mmPepMap™ Neo Trap Cartridge (part number 174500)
174502Nano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings5 mmPepMap™ Neo Trap Cartridge (part number 174500)
Catalog number 174501
Price (USD)/ Pack of 2
431.70
Add to cart
Description:
PEEK Tubing with nanoViper™ Fittings
Length (Metric):
100 mm
For Use With:
PepMap™ Neo Trap Cartridge (part number 174500)
Price (USD)/ Pack of 2
431.70
Add to cart
PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Catalog number174501
Price (USD)/ Pack of 2
431.70
-
Add to cart

Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.

See Viper and nanoViper resources

Specifications
Diameter (Metric)30 μm
For Use WithPepMap™ Neo Trap Cartridge (part number 174500)
IncludesTwo PEEK Tubes with nanoViper™ Fittings
Length (Metric)100 mm
Max. Pressure22,000 psi (1500 bar)
Temperature60°C
Product LinePepMap Neo
MaterialPEEK
DescriptionPEEK Tubing with nanoViper™ Fittings
For Use With (Application)Trap and Elute, Peptide Mapping
Unit SizePack of 2